SP Wilmad-LabGlass DURAN® Filter Flask, with Hose Connection, SP Industries
Proveedor: SP Industries
DURAN®
LG-7802-100
93000-864EA 16692.27 MXN
93000-864
SP Wilmad-LabGlass DURAN® Filter Flask, with Hose Connection, SP Industries
Matraces Matraces de filtro
Duran® Clear, heavy wall filter flask with removable hose connection.
  • With neck diameter of 70 mm