CleanSolutions™ Spray, Akron Biotechnology
AK5230-1000
89508-888EA 5665.52 MXN
89508-888
CleanSolutions™ Spray, Akron Biotechnology
CleanSolutions™ spray is a disinfectant solution that is used for all cleanroom and laboratory surfaces, including incubators and workstations
  • Non-toxic and odor-free
  • Ideally suited for disinfecting surfaces in cleanrooms and cell culture laboratories
It prevents the contamination and growth of bacteria, mycoplasma, fungi and viruses.