J-KEM® Temperature Controller, Model 210/T, With Timer, Chemglass
Proveedor: Chemglass
CG-3202-01 CG-3202-100 CG-3202-02 CG-3202-03
80062-920EA 49555.8 MXN
80062-918 80062-922 10034-458 80062-920
J-KEM® Temperature Controller, Model 210/T, With Timer, Chemglass
Controladores Controladores de temperatura
This model is sufficient for heating mantles up to 5 L in size along with most laboratory ovens and hot plates
This model also features an additional 100 hour digital timer to turn heating ON or OFF when time has expired.
Controller has the ability to display temperature without heating. Control temperature to within ± 0.1°C of liquids from 1 µL to 5 L.
Información de suministro: Includes 1/8” dia. x 12” long PTFE coated thermocouple, 10' coiled extension cord, and a 14/20 PTFE thermocouple adapter.