Magid® Gemstone Sapphire Y31 Protective Eyewear
Proveedor: Magid
Y31BKAFC
69100-098CS 16977.95 MXN
69100-098
Magid® Gemstone Sapphire Y31 Protective Eyewear
Gafas
Gemstone Sapphire Plus protective glasses feature a clear polycarbonate lens and side shields that are both impact resistant and fog resistant.
  • Light weight black, nylon frame with adjustable tilt rachet temple
  • Brow guard, integral nose bridge for enhanced comfort
Certificaciones: Meets ANSI Z87.1-2010 High Impact Standards.