Kindest Kare® Hand Cleanser, STERIS®
110587
14223-904CS 6133.87 MXN
14223-904
Kindest Kare® Hand Cleanser, STERIS®
Kindest Kare® combines the latest technology in skin conditioning to produce a quick rinsing hand and body lotion
  • Contains protein and Merquat™ skin conditioners that lubricate and moisturizer the skin
  • Opaque white in color with an herbal scent
The surface-active agents in Kindest Kare® disperse hard water, reducing soap residue and speeding the handwashing process.