Temperature and Humidity Logger, Compact (Dickson)
Proveedor: Dickson
TK550
97043-469PK 40171.82 MXN
97043-469
Temperature and Humidity Logger, Compact (Dickson)
Registradores de datos
This 12-pack of coin-sized temperature and humidity data loggers provide a great per unit price making them ideal for mapping warehouses or large stability chambers
  • 1 °C accuracy
  • Easy to use
  • Two channels: Temperature and humidity
  • Response time: 10 minutes to 63% of Full Scale
  • Average battery life: 6 months (at 2 minute sample rate, ambient temperature)
Thanks to their small size they are ideal for shipping and transportation applications.
Información para pedidos: Supplied with three volt lithium battery (one per unit) and Quick Start Guide. Software is needed for operation but not included. It can be ordered separately. PC requirements: Microsoft Windows 98 or newer; 16 MB RAM.