Analytical Scoops, Aluminium, Ohaus
Proveedor: Ohaus
80850321
12589-026EA 1429.68 MXN
12589-026
Analytical Scoops, Aluminium, Ohaus
Palas
These small analytical scoops made up of aluminium are used with measuring balances.