National Zoom Stereo Microscope with Tablet
BTW1-420TH
470229-748EA 42352.2 MXN
470229-748
National Zoom Stereo Microscope with Tablet
Features top light with directional beam, bottom light with cool, longer-life fluorescent illumination.
  • 8" LCD Android tablet with 1280 x 800 screen resolution
  • WiFI, mini-HDMI, and micros SD card connections
Now with detachable 8 inch tablet, this stereo zoom digital bundle package offers the convenience and versatility of viewing a large picture at 10X magnification, then zooming continuously all the way to close-up detail at 40X magnification. Upright, unreversed three-dimensional image remains in focus throughout the zoom range.
Like the Moticam X, this WiFi digital tablet transmits live images to IOS or Android devices. Use it as a conventional microscope or to share live images using WiFi tablets, wireless laptops and HD-ready monitor/projectors through HDMI. Tablet includes pre-loaded Motic apps.