Stirrer Shafts, Polished, Chemglass
Proveedor: Chemglass
CG-2097-10 CG-2097-12 CG-2097-52 CG-2097-51 CG-2097-30 CG-2097-75 CG-2097-53 CG-2097-31 CG-2097-11 CG-2097-16 CG-2097-08 CG-2097-100 CG-2097-50
89029-050EA 17267.97 MXN
16003-330 16003-332 89029-050 16003-334 16003-324 16003-336 89029-052 16003-326 89029-054 16003-328 89029-056 10170-264 10170-262
Stirrer Shafts, Polished, Chemglass
Imánes de agitación Ejes de agitación
These precision ground and polished stirrer shaft are used with process reactors
The 16003-336 and 16003-328 are made from heavy wall tubing and have intends for agitator positioning, which are spaced for every 10" from the bottom of agitator.
Designed to utilize with PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8" wide flat.