SUPOR® Disc Filter for Aspiration/Injection, 0.2 μm, B.Braun
Proveedor: B. Braun
Supor®
415002
10847-782CS 6893.22 MXN
10847-782
SUPOR® Disc Filter for Aspiration/Injection, 0.2 μm, B.Braun
Filtros Discos de filtros
These disc filters are designed for bacteria retentive filtration of medication.
  • DEHP-free and latex-free