Whatman™ pH Indicator Strips, Color Bonded, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
2613-991 2614-991 2613-991
28297-426PK 917.24 MXN
28297-426 51350-100 CAX28297426GSK
Whatman™ pH Indicator Strips, Color Bonded, Whatman products (Cytiva)
Papeles de pruebas Tiras indicadoras de pH
Can be used for the rapid determination of pH values and has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
Individual plastic support strips carry four different segments of dye-impregnated indicator papers. The resulting combination of colour differences gives an extremely clear and accurate visual pH value. All the dyes are chemically bonded to the paper and cannot be leached into solution; problems associated with contamination of the sample and resultant anomalous readings are avoided.