Polyester Labels, GA International
Proveedor: GA International
FreezerBondz™
60907 60903 60906 60908
80090-412EA 3071.88 MXN
80090-414 89005-248 89005-244 80090-412
Polyester Labels, GA International
Etiquetas
FreezerBondz™ B-490 Polyester Labels can be applied to already frozen surfaces but must wrap back onto itself to stay adhered upon being re-stored in liquid nitrogen. The white polyester material has a permanent acrylic adhesive and compatible with thermal transfer printers.
  • Good print smudge resistance, solvent resistance
  • High/low temperature performance from −196 to 100 °C
  • It does not provide viewing window for liquid level inside vial
  • All-in-one cartridge includes label supply and R4300 series ribbon
These durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.