SpectraMax® iD5 Multi-Mode Microplate Reader - Five-Mode Hybrid Microplate Reader with Automatic NFC Filter Identification and Western Blot Capability, Molecular Devices
Proveedor: Molecular Devices
5061514 6590-0086 6590-0140 6590-0110 6590-0110 6590-0124 6590-0108 6590-0133 6590-0101 5061514 6590-0129 6590-0080 6590-0112 6590-0095 6590-0130 6590-0080 6590-0152 6590-0083 5056339 6590-0117 6590-0101 6590-0120 ID5-STD 6590-0082 6590-0114 6590-0106 6590-0132 6590-0134 6590-0122 6590-0087 6590-0118 6590-0084 6590-0092 6590-0137 6590-0095 6590-0115 6590-0123 6590-0094 6590-0136 6590-0089 6590-0112 6590-0086 6590-0124 6590-0137 6590-0091 ID5-INJ-UPG 6590-0133 6590-0136 6590-0138 6590-0140 6590-0096 6590-0153 6590-0118 6590-0116 6590-0113 6590-0092 6590-0109 6590-0088 6590-0135 6590-0135 6590-0085 6590-0106 6590-0138 6590-0097 6590-0083 6590-0116 6590-0105 6590-0099 6590-0111 6590-0104 6590-0132 6590-0098 6590-0129 6590-0115 6590-0108 0200-7030 6590-0123 6590-0094 6590-0153 6590-0117 6590-0103 6590-0105 6590-0085 6590-0131 6590-0127 6590-0141 6590-0120 6590-0093 6590-0097 6590-0102 6590-0107 6590-0119 6590-0126 6590-0103 6590-0082 6590-0104 6590-0130 6590-0098 6590-0096 6590-0145 6590-0142 6590-0146 6590-0093 6590-0109 6590-0089 6590-0128 6590-0134 6590-0139 6590-0139 6590-0100 6590-0090 6590-0127 6590-0126 6590-0121 ID5-INJ-UPG 6590-0141 6590-0100 0200-7030 6590-0128 6590-0146 6590-0113 6590-0125 6590-0099 6590-0091 6590-0087 6590-0111 6590-0152 6590-0107 5056339 6590-0114 6590-0090 6590-0088 6590-0122 6590-0145 6590-0119 6590-0125 6590-0131 6590-0121 6590-0102 6590-0081 ID5-STD 6590-0081 6590-0142 6590-0084
CA76175-504 76175-616 CA76175-502 76175-614 CA76175-468 CA76175-500 76175-612 CA76175-466 76175-578 CA76175-620 76175-610 76175-576 CA76175-584 76175-574 CA76175-582 76175-572 CA76175-580 76175-570 CA76175-618 76175-608 CA76175-616 CA76175-516 76175-506 CA76175-514 76175-504 CA76175-512 76175-502 CA76175-478 76175-468 CA76175-598 CA76175-510 CA76175-476 76175-500 76175-466 CA76175-596 76175-620 CA76175-474 CA76175-594 76175-584 CA76175-472 CA76175-592 76175-582 CA76175-470 76175-580 CA76175-508 76175-618 CA76175-506 76175-518 76175-516 76175-514 CA76175-488 76175-512 76175-478 CA76175-520 76175-510 76175-598 CA76175-486 76175-476 76175-596 CA76175-484 76175-474 CA76175-482 76175-594 76175-472 CA76175-480 76175-592 76175-470 CA76175-518 76175-508 CA76175-538 CA76175-536 CA76175-534 CA76175-532 CA76175-498 CA76175-530 76175-488 76175-520 CA76175-496 76175-486 CA76175-494 76175-484 CA76175-492 76175-482 CA76175-490 76175-480 CA76175-548 76175-538 CA76175-546 76175-536 CA76175-544 76175-534 CA76175-542 76175-532 CA76175-540 76175-498 76175-530 76175-496 76175-494 76175-492 76175-490 CA76175-558 76175-548 CA76175-556 76175-546 CA76175-554 76175-544 CA76175-552 76175-542 CA76175-550 76175-540 CA76175-604 CA76175-602 CA76175-568 CA76175-600 76175-558 CA76175-566 76175-556 CA76175-564 76175-554 CA76175-288 CA76175-562 76175-552 CA76175-560 76175-550 76175-606 CA76175-614 76175-604 CA76175-612 76175-602 CA76175-578 CA76175-610 76175-568 CA76175-576 76175-600 76175-566 CA76175-574 76175-564 76175-288 CA76175-572 76175-562 CA76175-570 76175-560 CA76175-608 CA76175-606
SpectraMax® iD5 Multi-Mode Microplate Reader - Five-Mode Hybrid Microplate Reader with Automatic NFC Filter Identification and Western Blot Capability, Molecular Devices
Lectores de microplacas
The SpectraMax® iD5 Multi-Mode Microplate Reader is the complete laboratory solution to help you increase your research capabilities and comes with built-in absorbance, fluorescence, luminescence, time-resolved fluorescence (TRF), and tunable fluorescence polarization (FP) read modes
  • Easily set up protocols, run experiments, view tutorial videos, and control user access with the intuitive SoftMax Touch Software for the touchscreen
  • Automatically identify filter sets using NFC functionality, simplifying workflows and eliminating confusion
  • Perform western blot detection with your microplate reader
  • Push data to workstations using QuickSync, eliminating the need to retrieve data from the instrument
  • Detect assay signal in the best way using the superior hybrid monochromator- and filter-based optical system
  • Run injector assays with SmartInject™ Technology, ensuring complete reagent mixing and rapid signal development
A complete solution to answer all your research needs
The SpectraMax iD5 reader measures absorbance, fluorescence, luminescence, TRF, FP, and much more. The superior optical system includes a xenon flash lamp and features an ultra-cooled photomultiplier tube (PMT) that reduces background noise for excellent sensitivity and a wide dynamic range.
Featuring temperature control up to 66°C, linear, orbital, and double orbital shaking, a four-monochromator optical pathway with high efficiency gratings, option to use filters or a hybrid mix of monochromator and filterbased reads, well scanning up to a 20x20 read matrix, spectral scanning and detection of plate formats from 6- to 384-wells, the SpectraMax iD5 reader is the complete solution for all your research needs.
Expanded read mode capabilities
Equipped for high performance, the SpectraMax iD5 is HTRF®-certified, DLReady, and SpectraMax® DuoLuc™- certified. Microplate reader assays requiring infrared (IR) or red-shifted assays can be read with great sensitivity using the iD5 reader’s filter system. Specialized filters allow for IR fluorescence detection ranging from 10-40 times more sensitive than conventional monochromator detection reader. With the novel filter system, assays such as hydrogen peroxide detection assays and IRantibody-binding assays can be run confidently with Molecular Devices set of IR filters.
QuickSync data push technology
Reduce the amount of time you spend collecting data and increase your productivity. The SpectraMax iD5 reader allows you to interact with your data any way you want from anywhere you want. View your data quickly using the large touchscreen interface, export your data to a USB drive for analysis in the program of your choice, or analyze your data using the industry’s leading data acquisition and analysis tool, SoftMax Pro 7 Software. The SpectraMax iD5 reader also features network connectivity that allows you to walk away from the instrument to focus on additional research. Data is automatically delivered to any workstation on the same network, eliminating the need to physically retrieve data from the instrument.
Built on a foundation of excellence
For nearly 30 years, Molecular Devices has provided scientists with tools to expand the boundaries of their research. Our microplate readers are the industry’s most cited instruments and have empowered life science researchers to advance protein and cell biology, breaking the barriers to novel, landmark discoveries. The SpectraMax iD5 reader is built on the same foundation that has made our entire SpectraMax microplate reader product line among the most trusted in the industry
 In addition, the SpectraMax iD5 can be expanded to include bottom-read luminescence, TR-FRET, HTRF®, BRET, dual luciferase reporter assays with injectors, and western blot detection. 