Avantor® Alltima C18, HPLC Columns
Proveedor: VWR
Alltima®
AL-5C18-200D 81382 AL-10C18-250DF 970203 AL-3C18-250DF AL-10C18-300DF 96080/N 88334 50605 81142 AL-5C18-250DF 88051 AL-5C18-10CP2 88308 81388 AL-5C18-150DF 43851 AL-10C18-300DE 96680/N 88053 88057 81143 AL-3C18-100SP7 88059 96080SP/N 81106 81383 88383 88888 88353 88055 88307 96681/N 81105 88056 88052 AL-5C18-300D 81140 88335 81102 AL-10C18-150DE 5174010/N AL-3C18-250D AL-5C18-75D 43852 11350 81412 AL-3C18-75D 43853 AL-5C18-125D 88352 88370 88889 11543 AL-5C18-300DF AL-5C18-150DE AL-5C18-125DF 88371 AL-5C18-300DE 50603 81387 43803 88063 11541 88302
75797-230EA 30693.12 MXN
76398-426 76398-424 76398-422 76398-420 75797-226 75797-228 75830-846 75830-844 75797-230 75797-196 75798-824 75797-198 75798-822 75798-820 75797-190 75797-192 76398-418 75798-828 75797-194 76398-416 76398-414 76398-412 76398-410 75797-186 75813-078 75797-188 75797-220 75797-222 75797-224 75797-180 75797-182 75798-818 75797-184 76398-400 75813-070 75797-204 75813-072 75797-206 75813-074 75813-076 75797-174 75813-068 75830-796 75797-210 75797-176 75797-212 75797-178 75797-214 75797-172 76398-398 76398-430 76398-396 76398-394 76398-392 76398-390 75798-834 75797-168 75798-832 75830-384 75797-202 75798-830 75830-382 75831-456 75831-458 76398-428
Avantor® Alltima C18, HPLC Columns
Columnas de cromatografía
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
  • Base deactivated silica phases
  • Stable bonding for long column lifetime
  • Multiple selectivity options
  • USP L1
Classic reversed phase retention and selectivity. High quality hydrophobic general purpose C18.