VWR® Silica Gel Desiccants
VWRC83000.290 VWRC83001.290 VWRC83000.360
85007-082EA 3278.37 MXN
85007-082 85007-080 85007-084
VWR® Silica Gel Desiccants
Non toxic desiccants, for adsorbing moisture from the atmosphere and gas streams, which contain iron salts as a color indicator
  • Water adsorption capacity at 50% RH is a minimum of 23%
  • Loss on drying at 145 °C is less than 2%
  • At concentration of 10% in water, pH is from 1.5 to 5
  • Regeneration can be achieved by heating to 150 °C in a drying oven
The dry material has an intense orange color and changes to a pale beige/white color on adsorption of water.