Rotors for VWR® Laboratory Centrifuges
Proveedor: VWR, part of Avantor
VWR®
C3303-15-A57 C3303-50-A15 C3303-50-A57 C3303-2415 C3303-0450H C3303-0615H C3303-1810 C3303-1215 C3303-2410 C3303-0650
76181-194EA 6758.85 MXN
76181-194 76181-192 76181-198 76181-196 76181-202 76180-956 76181-200 76181-206 76180-952 76457-572
Rotors for VWR® Laboratory Centrifuges
Centrífugas Rotores
Swing out or angled rotors for separation.
  • Rotors for horizontal or angled separation
  • Accepts smaller tubes with the use of adapters
Swing out rotors: The 4×50 ml horizontal rotor and the 6×15 ml horizontal rotor of the VWR Laboratory Centrifuge are the perfect accessories for horizontal separation of 50 ml tubes and 15 ml tubes respectively. These rotors will also accept smaller tubes with the use of optional adapters.
Angled rotors: Rotors for angled separation include 18×10 ml, 24×10 ml, 12×15 ml, 24×15 ml and 6×50 ml. These rotors will also accept smaller tubes with the use of optional adapters.