Sample Vials, Amber, Phenolic Caps, Kimble Chase
Proveedor: DWK Life Sciences
60911B-12 60911B-1940 60911B-1528 60911B-10 60911B-2 60911B-5 60911B-1
89201-832CS 4016.95 MXN
89201-828 89201-820 89201-830 89201-824 89201-834 89201-822 89201-832
Sample Vials, Amber, Phenolic Caps, Kimble Chase
Viales
Vials manufactured from 51 expansion borosilicate amber glass conforming to USP Type I, with attached black phenolic caps and PTFE-faced white rubber liners.
Envase: Packaged in corrugated trays with partitions.