DEC-GLASS® Glass Cleaner, Veltek
Proveedor: Veltek
DEC-GLASS®
DG-03-16Z
14003-296CS 8008 MXN
14003-296
DEC-GLASS® Glass Cleaner, Veltek
Soluciones de limpieza Detergentes y Limpiadores de Superficie
DEC-GLASS® was developed for use in cleaning rotation cycles to remove residue left behind from disinfecting agents on glass or plexiglas surfaces.
  • Sterile and formulated with water for injection
  • Designed for use pharmaceutical and biotechnology operations