Disposable Magnetic Pad, CARDINAL HEALTH
Proveedor: Cardinal
200-16B
PH200-16ACS 11562.57 MXN
PH200-16A
Disposable Magnetic Pad, CARDINAL HEALTH
Suministros de primeros auxilios Apósitos quirúrgicos
Disposable magnetic pad covered with antistatic polyethylene.
It has lightweight drapes have multiple raised magnetic elements on a nonslip foam back. Allow positive instrument control adjacent to the surgical site.