Sucrose Solutions for Refractometers, Atago
Proveedor: Atago
RE-110020 RE-110040 RE-111001 RE-110060
97054-180EA 1654.97 MXN
97054-180 10859-884 10859-886 10859-930
Sucrose Solutions for Refractometers, Atago
Normativas Estándar para índice de refracción
Sucrose solutions are used for confirming the Brix measurements of refractometers.
  • Standard solutions have a shelf life of 6 weeks
  • High accuracy solutions have a shelf life of 10 days