Tamer Tabs Series 500, Walter Stern
Proveedor: Walter Stern
503
26398-004EA 181.02 MXN
26398-004
Tamer Tabs Series 500, Walter Stern
Piedras de ebullición
Tabs are highly effective in controlling distillation of liquids with low boiling points.
  • Made of alumina compound-porous granules
  • Melting point: 2040 °C
  • Non-reactive
Good in all boiling operations, except those with strong alkalis, or concentrated sulfuric acid.