Mini Economy Safety Glasses
Proveedor: DR INSTRUMENTS INC. SE
DR73802 DR70702 DR73801
470303-228CS 1561.19 MXN
470303-228 470303-230 470303-232
Mini Economy Safety Glasses
Gafas
Perfect for elementary students with small faces.
  • Available in three frame options
  • Perfect fit for small faces
  • Latex free
These anti-scratch, UV absorbing polycarbonate lens safety glasses provide outstanding impact strength and optical clarity for any laboratory application. The small size is perfect for younger students where safety and protection are imperative.