CleanMax® Clean and Sterile Disposable Hoods, Lakeland Industries
Proveedor: Lakeland Industries
CTL713CS
76193-232CS 14186.4 MXN
76193-232
CleanMax® Clean and Sterile Disposable Hoods, Lakeland Industries
Prendas desechables
Clean and Sterile disposable hoods are individually double-bagged and folded to promote easier gowning with less excursions.
  • Clean and sterile in silicone-free environment
  • Latex-free
  • Covers shoulders
  • One size
  • Ties to customize fit
  • Individually double-bagged
  • Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certificaciones: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.