Vaplock™ Chemical Exhaust Filter Adapters, Cole-Parmer
Proveedor: Cole Parmer
12019-88 12019-97 12019-86 12019-94 12019-89 12019-90 12019-99 12019-98 12019-92 12019-85 12019-87 12020-00 12019-96 12019-93 12019-91
76287-486EA 1439.22 MXN
76287-672 76287-486 76287-494 76287-670 76287-484 76287-492 76287-482 76287-490 76287-480 76287-676 76287-666 76287-710 76287-674 76287-488 76287-664
Vaplock™ Chemical Exhaust Filter Adapters, Cole-Parmer
Racores y conectores
Exhaust filters may be attached to various point-source emissions by the use of Vaplock™ adapters.
Filter Adapters offer versatile configurations.
Please note that options for justrite safety containers are made to fit either stainless steel or polypropylene quick disconnects, but are not interchangeable.
Vaplock™ EF-100 and EF-200 exhaust filters may be attached to various point-source emissions by use of the appropriate adapter. Each filter includes a 1/4" female NPT port; select from the list below to adapt filters to a typical system, or inquire for assistance with special requirements. All adapters are reusable.