Dual-Wire Fabric Wristband, SCS, Desco Industries
Proveedor: Desco Industries
2360R 2371 2370 2370R 2360 2371 2368
76238-062EA 1445.32 MXN
76238-064 76238-065 76243-338 76238-060 76238-062 76239-604 76311-422
Dual-Wire Fabric Wristband, SCS, Desco Industries
Tomas de tierra Muñequeras antiestáticas
The Dual-Wire Fabric Wristband is available in one size that adjusts to almost any size wrist for reliable protection.
  • Adjustable, elastic nylon material with conductive woven thread
  • Fits most wrist sizes to provide reliable protection
  • Dual-Wire conductor provides redundancy
  • Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
The wristband provides full conductivity, comfort, and reliability, while ensuring continuous drain of static charge when used with SCS 2300 Dual-Wire Coil Cords (sold separately). Made to be used in conjunction with all SCS Workstation Monitors.
Certificaciones: Meets requirements of ANSI/ESD S20.20 and ANSI/ESD S1.1.