Pioneer® Precision Balances
Supplier: OHAUS SCALE CORP
Certificates
About this item
Affordable balance to achieve reliable results. The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings.
- Features cast metal lower housing, sub-pan and stainless steel weighing pan
- PX is durably constructed for versatile, long-term use
- Second line display for additional information or guidance
- Static removal bar for convenient grounding
- USB connectivity
The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
Specifications
- Catalog No:
- 76289-408
- 76289-414
- 76289-416
- Communication interface:
- RS232; USB
- RS232; USB
- RS232; USB
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
Specifications
Analytical Balances
Product Information
- Catalog #Supplier #ModelWeighing CapacityLinearityPan/Platform SizeReadabilityStabilization TimeRepeatabiltyAvailabilityPrice
- 76289-40830430053PX224 (Internal Calibration)220 g±0.0002 gØ 90 mm0.1 mg3 s0.0001 gLog in for availabilityEach (1UN)$99,965.54Specifications:CSA CertifiedIrregular Voltage
More
Specifications
Environmentally PreferableCommunication interfaceRS232; USBVoltage100 - 240 VCSA CertifiedIrregular Voltage - 76289-41430429849PX323/E320 g±0.002 gØ 120 mm0.001 g2 s0.001 gLog in for availabilityEach (1UN)$43,874.42Specifications:CSA CertifiedIrregular Voltage
More
Specifications
Environmentally PreferableCommunication interfaceRS232; USBVoltage100 - 240 VCSA CertifiedIrregular Voltage - 76289-41630429840PX323 (Internal Calibration)320 g±0.002 gØ 120 mm0.001 g2 s0.001 gLog in for availabilityEach (1UN)$53,127.82Specifications:CSA CertifiedIrregular Voltage
More
Specifications
Environmentally PreferableCommunication interfaceRS232; USBVoltage100 - 240 VCSA CertifiedIrregular Voltage