Luer Lock Unassembled Manual Syringes, Jensen Global
Proveedor: JENSEN GLOBAL INC
JGM5LL-20
76243-116EA 698.78 MXN
76243-116
Luer Lock Unassembled Manual Syringes, Jensen Global
Jeringas Jeringas de uso múltiple
These syringes come unassembled without any calibrations and are sold in packages with the barrel, stopper, and plunger.