Inserts for 8-425 Glass Screw-Thread Vials, Thermo Scientific
Proveedor: Thermo Scientific
C4012-530 C4012-530P C4012-S530
66011-200PK 3295.17 MXN
66011-200 66030-462 66065-264
Inserts for 8-425 Glass Screw-Thread Vials, Thermo Scientific
Viales
Polyspring inserts are self-aligning and provide a cushion against needle contact.
These clear glass or polypropylene inserts for 8 mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125 to 200 μl.