Inserts for Target DP™ 9-425 Glass Screw-Thread Vials, Thermo Scientific
Proveedor: Thermo Scientific
C4010-629 C4010-K630 C4010-S629 C4010-629L C4011-631P C4011-S631 C4011-631 C4010-630 C4010-630P C4010-630TS C4010-S630 C4010-627L
66030-902PK 2889.33 MXN
66030-900 66065-336 66064-184 66064-168 66064-226 66064-258 66030-902 66030-898 66030-904 66065-236 66064-186 66064-166
Inserts for Target DP™ 9-425 Glass Screw-Thread Vials, Thermo Scientific
Viales Insertos para viales de cromatografía
The polyspring microseal inserts self-align and provide a cushion against needle contact.
Precision point inserts minimize residual sample loss. Pulled point inserts provide an economical choice for noncritical applications.