Septa for 13-425 Screw-Thread Vials, Thermo Scientific
Proveedor: Thermo Scientific
C4015-30 C4015-45 C4015-61 C4015-10 C4015-60 C4015-55 C4015-40
66064-318PK 1558.09 MXN
66030-390 66064-310 66065-226 66030-388 66064-318 66064-306 66030-962
Septa for 13-425 Screw-Thread Vials, Thermo Scientific
Septos
Autosampler vial septa, 13 mm for use with 13-425 threaded caps or 13 mm snap/crimp caps.
  • PTFE disk septa are chemically inert
  • PFTE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
Septa are available in PTFE/ red rubber, PTFE/silicone or red PTFE/white silicone/red PTFE to suit a variety of applications.