Finger Cot, 8C, Quality Rubber Products
Proveedor: QRP
8CL
76243-044CS 30939.22 MXN
76243-044
Finger Cot, 8C, Quality Rubber Products
Protectores de dedos Protectores para dedos de látex
The QRP 8C static dissipative latex finger cots is recommended for use with static sensitive devices.
  • Unique, stand-up pouch for easy product identification
  • Packaged for ISO 5 (Class 100) applications
  • Average surface resistance of 5×107 ohms per square unit
  • For use with Class I and II static sensitive devices
They are widely used for ESD sensitive areas, defense, avionics, marine electronics, consumer electronics, IC industries, wire bonding, repair stations, die bonding, laser diodes and field repair kits.
Certificaciones: Compliance to ASTM and IES-RP-CC 005.3 standards.
Precaución: Finger cots contain natural rubber latex, which may cause allergic reactions.