Syringe Barrels and Piston Sets, EFD
Proveedor: Nordson EFD
7012153 7012096 7012114 7012091 7012074 7360577 7012075 7012085 7012136 7012118 7012149
76238-214BX 1168.35 MXN
76247-646 76241-722 76267-510 76245-646 76241-166 76238-214 76241-010 76247-468 76245-488 76252-438 76243-808
Syringe Barrels and Piston Sets, EFD
Émbolos de jeringa
Precision fit between syringe and piston ensures consistent fluid deposits.
  • Silicone-free
  • Package labels include lot numbers for process control and traceability
Wiper piston improves fluid control, keeps fluids from dripping and eliminates waste by wiping the syringe wall clean.
Sets are available in clear for most fluids, transparent amber for UV and light-sensitive materials (up to 550 nm) and opaque black for complete light blockage.
Información de suministro: Each box contains one resealable bag of syringes and SmoothFlow™ pistons.
Envase: Dust-free packaging.