Order Entry
CHR® Tapes, K Series, Adhesive, Saint-Gobain
CHR® Tapes, K Series, Adhesive, Saint-Gobain
K109 Series
  •  
    Color
    Adhesive material
    Width
    Thickness
    Elongation
    Tensile Strength
  • Acrylic adhesive type
    Acrylic adhesive type
    76243-356
    CHR-K109-.405
    Amber
    Pressure sensitive
    1.02 cm (0.405")
    0.0089 cm
    75% at break
    88 N/cm
    Each (1UN)
    Each (1UN) - $1,927.55
    $1,927.55
    Each (1UN)
       $1,927.55
    Specifications
    Environmentally Preferable
K350 Series
  •  
    Color
    Adhesive material
    Width
    Thickness
    Elongation
    Tensile Strength
  • Silicone adhesive type
    Silicone adhesive type
    76243-348
    CHR-K350-.500
    Amber
    Pressure sensitive
    1.2 cm (0.500")
    0.0089 cm
    75% at break
    88 N/cm
    Each (1UN)
    Each (1UN) - $1,668.86
    $1,668.86
    Each (1UN)
       $1,668.86
    Specifications
    Environmentally Preferable
  • Silicone adhesive type
    Silicone adhesive type
    76243-350
    CHR-K350-1.2
    Amber
    Pressure sensitive
    3 cm (1.2")
    0.0089 cm
    75% at break
    88 N/cm
    Each (1UN)
    Each (1UN) - $3,926.14
    $3,926.14
    Each (1UN)
       $3,926.14