Soldering Cartridges, STTC, OK International
Proveedor: OK International
STTC-113 STTC-117-PK STTC-125P STTC-144 STTC-137 STTC-122 STTC-026 STTC-101 STTC-147P STTC-022 STTC-107 STTC-142 STTC-037-PK STTC-025 STTC-136 STTC-126 STTC-101P STTC-146 STTC-138 STTC-135 STTC-840 STTC-838 STTC-042 STTC-147 STTC-145 STTC-114 STTC-138P STTC-125-PK STTC-112 STTC-099 STTC-143 STTC-137P STTC-136P STTC-145P STTC-047 STTC-140
76242-570EA 1300.48 MXN
76242-756 76242-758 76242-972 76244-986 76242-970 76242-570 76241-518 76241-516 76241-538 76241-536 76241-514 76241-534 76241-532 76241-530 76241-692 76241-690 76242-768 76242-764 76242-622 76242-760 76242-564 76242-184 76241-528 76241-526 76277-150 76243-378 76241-524 76241-522 76241-688 76241-520 76241-542 76241-686 76241-540 76241-684 76242-618 76254-262
Soldering Cartridges, STTC, OK International
Soldadura Elementos para soldar
Compatible with MX-500 and MX-5000 Systems / MX-RM3E and MX-H1-AV hand-pieces.