High Temperature Polyimide Labels for Thermal Transfer Printers, Brady
Proveedor: Brady
UltraTemp
139871 139870 139860 132462 139837 139863 132469
76243-632EA 16035.36 MXN
76243-632 76246-658 76268-018 76277-266 76249-270 76242-068 76253-176
High Temperature Polyimide Labels for Thermal Transfer Printers, Brady
Etiquetas
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Resists high temperatures up to 572 °F (300 °C)
  • Survives on the top or bottom side of printed circuit boards through the solder process
Withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component preprocess labeling.
Certificaciones: Fulfills requirements for MIL-STD-202G Method 215K and SAE AS81531 Marking of Electrical Insulating Material when used with R4300 series ribbons.