Interchangeable Glass Syringe with Luer Lock Tip, Ace Glass Incorporated
Proveedor: Ace Glass
13681-02 13681-50 13681-05 13681-20 13681-30 13681-10
76312-824EA 1676.88 MXN
76312-826 76312-828 76312-822 76312-824 76312-830 76312-832
Interchangeable Glass Syringe with Luer Lock Tip, Ace Glass Incorporated
Jeringas Jeringas de uso múltiple
Interchangeable syringe featuring a Luer lock tip, permanently fused graduations, and a colored plunger end for easy and accurate dosage measurement.
  • Made of high-quality borosilicate glass
  • Reinforced flanges that are flat on two sides to prevent rolling
  • Marked for non-medical use; no medical license required
Manufactured to ensure the perfect mating of barrels and plungers, so any interchangeable plunger of a particular size will fit any interchangeable barrel of the same size.