VWR® Media Bottles, Square, PET, Sterile
Proveedor: VWR
VWR®
PET250-T PET500 PET1000-T PET500-T PET250 PET1000
76302-840CS 2092.77 MXN
76299-562 76302-840 76299-560 76302-838 76302-836 76299-564
VWR® Media Bottles, Square, PET, Sterile
Frascos Botellas de plástico
Square PET media bottles are designed for packaging, storing, or transporting powder and liquid samples.
  • Square design allows efficient use of storage space
  • PET resin is lightweight, break-resistant, and transparent
  • Bottles feature molded-in graduations
  • EB Sterilization Sterility Assurance Level (SAL) of 10⁻⁶ (ISO 11137)
  • Non-pyrogenic
VWR® Square PET Media Bottles are manufactured in a class 100000 cleanroom environment. Manufactured under EN ISO 13485:2016.