VWR® Freezer Storage Boxes, 81-Place, 50-Place and 100-Place
Proveedor: VWR
VWR®
5500-00-22AV1 5510-29-22AV1 5510-00-22AV1 5500-29-22AV1 5540-00-22AV1 5540-29-22AV1
76333-284CS 7374.96 MXN
76333-298 76333-288 76333-300 76333-284 76333-296 76333-286
VWR® Freezer Storage Boxes, 81-Place, 50-Place and 100-Place
Gradillas Gradillas de viales criogénicos
VWR® Freezer boxes are made of polypropylene and come with an alphanumerically labeled grid for identifying contents.
  • Boxes are autoclavable
  • Freezable to –90 °C (–130 °F)
The 81-place box is designed to fit into existing freezer storage racks, holds 1.5 or 2.0 ml microcentrifuge tubes, and includes an attached clear lid. The 50 and 100-place freezer boxes come with attached hinged lid and snap lock closure for versatility. They are designed to hold 1.5 and 2.0 ml tubes and some 1.2 ml cryo vials. Indented lids can be labeled.
Información de suministro: Assorted colors packages include one each in blue, green, orange, red, and yellow.