Aspirator
470320-974
- Description:Aspirator
- Cat. no.:470320-974
Strong plastic aspirator for vacuum filtration and other lab applications.
- Prevents flooding and back flow
Designed to prevent any flooding, back-flow, or turbulence. Includes 5.9" (15 cm) hose. The top nozzle has an outer diameter of 1.5 cm. The side nozzle can accommodate hoses from 0.5 to 1.0 cm inner diameter.