KIMBLE® Watch Glasses, DWK Life Sciences
Proveedor: DWK Life Sciences
KIMBLE®
72001-70 72001-90 72001-150 72001-80 72001-100 72001-50 72001-125
75797-132CS 425.73 MXN
75797-142 75797-130 75797-132 75797-134 75797-136 75797-138 75797-140
KIMBLE® Watch Glasses, DWK Life Sciences
Vidrios de relojes
Concave circular glasses are used to perform evaporations for liquids, to cover beakers and to hold solids while weighing.
  • Minimum thickness 1.2 mm
  • Diameter tolerance ±1 mm
  • Fire polished edge
Manufactured from 90 expansion soda-lime glass.
Certificaciones: Conforming to USP Type III requirements.