/
Mexico
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Ward's® Mini LED White-Light Box
470311-200
:
VWR International
Ward's® Mini LED White-Light Box
470311-200
:
VWR International
$4,847.25
Select uom
Unit of Measure
Each (1UN)
-
+
Description:
Mini light box
Dimensions:
Overall: 158×150×14 mm (6×6×0.5")
Work area: 125×100 mm (5×4")
Cat. no.:
470311-200