Cryo-Marker™ Dual Point Cryogenic Water-Resistant Markers, GA International
Proveedor: GA International
CryoMarker™
MP-1BL MP-1BR MP-1YE 807656002781 807656002804
76276-064EA 129.17 MXN
76276-064 76276-068 76276-078 76276-066 76276-076
Cryo-Marker™ Dual Point Cryogenic Water-Resistant Markers, GA International
Bolígrafos Rotuladores criogénicos
Dual point high-quality permanent ink marker.
  • Excellent performance on plastic, glass, metal, cloth, and other surfaces
  • Compatible with Lab-TAG™ Labels from GA International
  • Easy to handle for applications in bio-medical laboratories
  • Extra-Fine (0.3 mm) and fine (0.7 mm) tips; 2 pens in 1
  • Can write on Teflon surfaces
  • Low odor
Durable, heavy-duty, long lasting markers withstand rough treatment and provide delicate touch on various materials. Permanent and waterproof, all colors are equally permanent and lasting.