Gen-X S-1 Powder-Free Nitrile Examination Gloves, Smart Glove
Proveedor: VWR International
PS-249A-S PS-249A-XL PS-249A-L PS-249A-M
76449-370CS 8580 MXN
76449-370 76449-364 76449-368 76449-366
Gen-X S-1 Powder-Free Nitrile Examination Gloves, Smart Glove
Guantes
Gen-X S-series nitrile gloves provide a comfortable and soft feel to the wearer. These gloves are designed and produced with low modulus and high elongation, reducing wearer fatigue. They are suitable for medical personnel, veterinary operations, pharmaceutical, and general hygiene use.
  • Finger thickness: 3 mil
  • Medical grade
  • Finger textured with beaded cuffs
  • Phthalate-free
Gloves are manufactured with a special blend of nitrile to produce a soft material that enhances comfort and prevents hand fatigue.
Certificaciones: ASTM D6319. Manufactured in compliance with ISO 9001, ISO 13485, and MDSAP.
Envase: Packed 100 gloves pers dispenser box, 10 boxes per case.
Precaución: Keep storage area cool, dry, and dust-free, avoid ventilation and storage close to photocopy equipment. Copper ions discolor the glove. Protect gloves against ultraviolet light sources, such as sunlight and oxidizing agents. Storage above 38 °C (100 °F) will lead to accelerated aging and should be avoided under any circumstances.