IUPAC Certified pH Buffer Standards, Hach
Proveedor: Hach
S11M008 S11M001 S11M004 S11M003 S11M007 S11M005 S11M006 S11M002
76492-562EA 1875.23 MXN
76492-572 76492-560 76492-562 76492-564 76492-566 76492-568 76492-570 11024-744
IUPAC Certified pH Buffer Standards, Hach
Tampones Tampones pH
Buffers traceable to IUPAC/NIST pH scale.
  • Guaranteed shelf-life thanks to premium packaging
  • Colourless pH buffer solution
Each standard is delivered in an airtight package ensuring safe storage. The nominal value of the pH standard is guaranteed until first opening. Certified value is guaranteed for two years from the date of issue of the certificate.