VWR® Standard Series Manual Defrost Laboratory Freezers with Natural Refrigerants
Proveedor: VWR, part of Avantor
VWR®
HCMFS-14 HCMFS-20 HCMFS-17
75836-238EA 135803.29 MXN
75836-238 76177-832 76498-984
VWR® Standard Series Manual Defrost Laboratory Freezers with Natural Refrigerants
Congeladores Congeladores de laboratorio/aplicaciónes generales
These efficient and economical freezers provides quality cold storage in laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant (R290)
These manual defrost freezers offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness. These freezers are equipped with keyed door locks, interior shelving, probe access, and microprocessor temperature controllers.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. The freezers run on natural, hydrocarbon refrigerant for environmental health and economic efficiency. Ideal for use in laboratory.
Certificaciones: UL or ETL Listed.