VWR® Plus Laboratory Refrigerators, Stainless Steel
Proveedor: VWR
VWR®
VWR-HC-SSP-49G VWR-HC-SSP-23G VWR-HC-SSP-72 VWR-HC-SSP-49 VWR-HC-SSP-72G VWR-HC-SSP-23
76514-208EA 115200 MXN
76514-210 76514-212 76514-206 76514-208 76514-202 76514-204
VWR® Plus Laboratory Refrigerators, Stainless Steel
Refrigeradores
These VWR® Plus Series stainless steel refrigerators feature a multi-functional design for laboratory use.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Forced draft air circulation
  • Natural, hydrocarbon refrigerant
These VWR® refrigerators utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. All models are equipped with probe access ports, adjustable shelving, and interior lighting. Units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Certificaciones: ETL Listed.