Order Entry
VWR® Mega Star 4.0 General Purpose 4 L Benchtop Centrifuges
VWR® Mega Star 4.0 General Purpose 4 L Benchtop Centrifuges
undefined
VWR® Mega Star 4.0 General Purpose 4 L Benchtop Centrifuges
  • Catalog No:
  • 76468-136
  • 76468-138
  • 76519-286
  • 76519-284
  • Max. speed:
  • 15200 rpm (depending on rotor)
  • 15200 rpm (depending on rotor)
  • 15200 rpm (depending on rotor)
  • 15200 rpm (depending on rotor)
  • Max. RCF:
  • 25830×g (depending on rotor)
  • 25830×g (depending on rotor)
  • 25830×g (depending on rotor)
  • 25830×g (depending on rotor)
  • Voltage:
  • 120 V
  • 120 V
  • 120 V
  • 120 V
  • Height:
  • Lid open: 86.1 cm (33.9")
    Lid closed: 36.1 cm (14.2")
  • Ld open: 86.1 cm (33.9")
    Lid closed: 36.2 cm (14.3")
  • Ld open: 86.1 cm (33.9")
    Lid closed: 36.2 cm (14.3")
  • Lid open: 86.1 cm (33.9")
    Lid closed: 36.1 cm (14.2")
  • Depth:
  • 66 cm (26")
  • 66 cm (26")
  • 66 cm (26")
  • 66 cm (26")
  • Width:
  • 74.6 cm (29.4")
  • 56.6 cm (22.3")
  • 56.6 cm (22.3")
  • 74.6 cm (29.4")
Centrifuges
  •  
    Capacity
    Temperature Range
    Weight
  • VWR® Mega Star 4.0 general purpose 4 L benchtop centrifuge, ventilated, 120 V (centrifuge only)
    VWR® Mega Star 4.0 general purpose 4 L benchtop centrifuge, ventilated, 120 V (centrifuge only)
    76468-138
    4.0 L
    89 kg (196 lbs.)
    Each (1UN)
    Each (1UN) - $208,930.00
    $208,930.00
    Each (1UN)
       $208,930.00
    Specifications
    Environmentally Preferable
    Max. speed
    15200 rpm (depending on rotor)
    Max. RCF
    25830×g (depending on rotor)
    Voltage
    120 V
    Height
    Ld open: 86.1 cm (33.9")
    Lid closed: 36.2 cm (14.3")
    Depth
    66 cm (26")
    Width
    56.6 cm (22.3")
  • VWR® Mega Star 4.0 R general purpose 4 L benchtop centrifuge, refrigerated, 120 V (centrifuge only)
    VWR® Mega Star 4.0 R general purpose 4 L benchtop centrifuge, refrigerated, 120 V (centrifuge only)
    76468-136
    4.0 L
    −10...+40 °C
    117 kg (258 lbs.)
    Each (1UN)
    Each (1UN) - $343,665.36
    $343,665.36
    Each (1UN)
       $343,665.36
    Specifications
    Environmentally Preferable
    Max. speed
    15200 rpm (depending on rotor)
    Max. RCF
    25830×g (depending on rotor)
    Voltage
    120 V
    Height
    Lid open: 86.1 cm (33.9")
    Lid closed: 36.1 cm (14.2")
    Depth
    66 cm (26")
    Width
    74.6 cm (29.4")
Centrifuge packages
  •  
    Capacity
    Temperature Range
    Weight
  • VWR® Mega Star 4.0 general purpose 4 L benchtop centrifuge tissue culture package, ventilated, 120 V (tissue culture packages includes TX1000 rotor, set of 4×1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters)
    VWR® Mega Star 4.0 general purpose 4 L benchtop centrifuge tissue culture package, ventilated, 120 V (tissue culture packages includes TX1000 rotor, set of 4×1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters)
    76519-286
    4.0 L
    89 kg (196 lbs.)
    Each (1UN)
    Each (1UN) - $332,330.08
    $332,330.08
    Each (1UN)
       $332,330.08
    Specifications
    Environmentally Preferable
    Max. speed
    15200 rpm (depending on rotor)
    Max. RCF
    25830×g (depending on rotor)
    Voltage
    120 V
    Height
    Ld open: 86.1 cm (33.9")
    Lid closed: 36.2 cm (14.3")
    Depth
    66 cm (26")
    Width
    56.6 cm (22.3")
  • VWR® Mega Star 4.0 R general purpose 4 L benchtop centrifuge tissue culture package, refrigerated, 120 V (tissue culture packages includes TX1000 rotor, set of 4×1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters)
    VWR® Mega Star 4.0 R general purpose 4 L benchtop centrifuge tissue culture package, refrigerated, 120 V (tissue culture packages includes TX1000 rotor, set of 4×1000 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 10×50 ml conical tube adapters, set of four 24×15 ml conical tube adapters)
    76519-284
    4.0 L
    −10...+40 °C
    117 kg (258 lbs.)
    Each (1UN)
    Each (1UN) - $480,307.13
    $480,307.13
    Each (1UN)
       $480,307.13
    Specifications
    Environmentally Preferable
    Max. speed
    15200 rpm (depending on rotor)
    Max. RCF
    25830×g (depending on rotor)
    Voltage
    120 V
    Height
    Lid open: 86.1 cm (33.9")
    Lid closed: 36.1 cm (14.2")
    Depth
    66 cm (26")
    Width
    74.6 cm (29.4")