Masterflex® L/S® Pump Tubing, Gore® STA-PURE® PFL-Series, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
ZA12FE-MFLX CA12FE-MFLX BB14FE-MFLX CB14FE-MFLX BA12FE-MFLX AB14FE-MFLX AA12FE-MFLX DB24FE-MFLX YA12FE-MFLX
MFLX96242-17EA 21905 MXN
MFLX96242-17 MFLX96242-16 MFLX96242-18 MFLX96242-24 MFLX96242-35 MFLX96242-15 MFLX96242-25 MFLX96242-36 MFLX96242-14
Masterflex® L/S® Pump Tubing, Gore® STA-PURE® PFL-Series, Avantor®
Tuberías Tubería de bomba peristáltica
High-performance pump tubing for challenging applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Custom extruded to fit Masterflex® pumps
  • Lot-to-lot consistency provides superior accuracy and repeatability
Applications: Chemical and pharmaceutical applications
Characteristics: Enhanced chemical resistance, including aggressive organic solvents, spallation-free, low gas permeability, opaque white
Gore® STA-PURE® PFL-Series Pump Tubing is your ideal choice for high purity or long-running, critical pharmaceutical and biopharmaceutical processing applications. The patented composite (perfluoroelastomer) and ePTFE (polytetrafluoroethylene), results in a tube that offers high flex durability for greater resistance to rupture—even at applications up to 60 psig. Long-term pump testing proves that Gore® STA-PURE® PFL-Series Pump Tubing lasts hundreds of hours with consistent and repeatable flow rates resulting in one saving time and reducing maintenance by having fewer tubing changes.
If one is looking for a flow stable / process stable tubing that withstands CIP (clean-in-place) or SIP (steam-in-place) cleaning—look no further than this tubing!
Gore® STA-PURE® Pump tubing - Series PFL is available as single tube elements. Recommended for use in L/S® and I/P® standard and Easy-Load® pump heads, and L/S® Easy-Load® II pump heads.
NOTE: There is an engineering questionnaire vetting application specific details before the order can be processed.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.