Order Entry
Masterflex® Single-Use Stainless Steel Turbine Flow Sensors, Avantor®
Masterflex® Single-Use Stainless Steel Turbine Flow Sensors, Avantor®
Supplier:  VWR International
Certificates
undefined
Masterflex® Single-Use Stainless Steel Turbine Flow Sensors, Avantor®
Supplier:  VWR International
Specifications
  • Model:
  • 24507-38
  • 24507-40
  • 24507-39
  • Accuracy:
  • 1% of reading
  • 1% of reading
  • 1% of reading
  • Max. pressure:
  • 1450 psi
  • 2900 psi
  • 2900 psi
  • Min. operating temperature:
  • –20 °C (–4 °F)
  • –20 °C (–4 °F)
  • –20 °C (–4 °F)
  • Max. operating temperature:
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • 80 °C (176 °F)
  • Viscosity:
  • 0.8 - 10 cSt
  • 0.8 - 10 cSt
  • 0.8 - 10 cSt
  • Repeatability:
  • <0.15%
  • <0.15%
  • <0.15%
  • Float material:
  • Ruby
  • Ruby
  • Ruby
  • O-Ring material:
  • EPDM
  • EPDM
  • EPDM
  • Wetted materials:
  • 316 SS, PVDF, and ruby sensor with EPDM o-ring
  • 316 SS, PFA, and ruby sensor with EPDM o-ring
  • 316 SS, PVDF, and ruby sensor with EPDM o-ring
  • Certification:
  • Single-Point Calibration with Certificate
  • Single-Point Calibration with Certificate
  • Single-Point Calibration with Certificate
  • Power:
  • 5 - 24 VDC
  • 5 - 24 VDC
  • 5 - 24 VDC
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Information
  • Catalog #
    Fluid Type
    Maximum Flow Rate
    Min. flow rate
    Output
    Fitting Material
    Housing Material
    Process Connection
    Availability
    Price
  • Turbine Flow Sensor, 316L Stainless Steel, 0,03 to 2 L/min, ¹/₄" NPT(M), with Single-Point Calibration Certificate
    Turbine Flow Sensor, 316L Stainless Steel, 0,03 to 2 L/min, ¹/₄" NPT(M), with Single-Point Calibration Certificate
    MFLX24507-38
    Conductive or non-conductive fluids with no lenticular impurities
    2 L/min
    0.03 L/min
    One 5 to 24 volt square wave pulse
    Stainless steel 316
    316 stainless steel and PVDF
    0.25" NPT(M)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $14,240.00
    $14,240.00
    Each (1UN)
       $14,240.00
    Specifications:
    1 year warranty
    More
    Accessories
    Specifications
    Model
    24507-38
    Environmentally Preferable
    Accuracy
    1% of reading
    Max. pressure
    1450 psi
    Min. operating temperature
    –20 °C (–4 °F)
    Max. operating temperature
    80 °C (176 °F)
    Viscosity
    0.8 - 10 cSt
    Repeatability
    <0.15%
    Float material
    Ruby
    O-Ring material
    EPDM
    Wetted materials
    316 SS, PVDF, and ruby sensor with EPDM o-ring
    Certification
    Single-Point Calibration with Certificate
    Power
    5 - 24 VDC
    Warranty
    1 year
    1 year warranty
  • Turbine Flow Sensor, 316L Stainless Steel, 0,5 to 20 L/min, ³/₈" NPT(M), with Single-Point Calibration Certificate
    Turbine Flow Sensor, 316L Stainless Steel, 0,5 to 20 L/min, ³/₈" NPT(M), with Single-Point Calibration Certificate
    MFLX24507-39
    Conductive or non-conductive fluids with no lenticular impurities
    20 L/min
    0.5 L/min
    One 5 to 24 volt square wave pulse
    Stainless steel 316
    316 stainless steel and PVDF
    0.375" NPT(M)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $14,240.00
    $14,240.00
    Each (1UN)
       $14,240.00
    Specifications:
    1 year warranty
    More
    Accessories
    Specifications
    Model
    24507-39
    Environmentally Preferable
    Accuracy
    1% of reading
    Max. pressure
    2900 psi
    Min. operating temperature
    –20 °C (–4 °F)
    Max. operating temperature
    80 °C (176 °F)
    Viscosity
    0.8 - 10 cSt
    Repeatability
    <0.15%
    Float material
    Ruby
    O-Ring material
    EPDM
    Wetted materials
    316 SS, PVDF, and ruby sensor with EPDM o-ring
    Certification
    Single-Point Calibration with Certificate
    Power
    5 - 24 VDC
    Warranty
    1 year
    1 year warranty
  • Turbine Flow Sensor, 316L Stainless Steel, 1,5 to 40 L/min, ¹/₂" NPT(M), with Single-Point Calibration Certificate
    Turbine Flow Sensor, 316L Stainless Steel, 1,5 to 40 L/min, ¹/₂" NPT(M), with Single-Point Calibration Certificate
    MFLX24507-40
    Conductive or non-conductive fluids with no lenticular impurities
    40 L/min
    1.5 L/min
    One 5 to 24 volt square wave pulse
    Stainless steel 316
    316 stainless steel and PFA
    0.5" NPT(M)
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $14,240.00
    $14,240.00
    Each (1UN)
       $14,240.00
    Specifications:
    1 year warranty
    More
    Accessories
    Specifications
    Model
    24507-40
    Environmentally Preferable
    Accuracy
    1% of reading
    Max. pressure
    2900 psi
    Min. operating temperature
    –20 °C (–4 °F)
    Max. operating temperature
    80 °C (176 °F)
    Viscosity
    0.8 - 10 cSt
    Repeatability
    <0.15%
    Float material
    Ruby
    O-Ring material
    EPDM
    Wetted materials
    316 SS, PFA, and ruby sensor with EPDM o-ring
    Certification
    Single-Point Calibration with Certificate
    Power
    5 - 24 VDC
    Warranty
    1 year
    1 year warranty