Masterflex® Union Fittings, Compression, Straight, Avantor®
Proveedor: Avantor Fluid Handling
Masterflex®
MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP MAKE-UP
MFLX06381-50EA 1353 MXN
MFLX06381-50 MFLX06381-60 MFLX30803-06 MFLX30803-07 MFLX30803-04 MFLX30803-05 MFLX06382-15 MFLX30805-04 MFLX30805-05 MFLX06382-14 MFLX30803-03 MFLX30805-02 MFLX06382-13 MFLX06382-12 MFLX30805-03 MFLX30805-00 MFLX30802-95 MFLX06382-11 MFLX06381-10 MFLX06381-20 MFLX30805-01 MFLX30802-94 MFLX06381-30 MFLX06381-40
Masterflex® Union Fittings, Compression, Straight, Avantor®
Racores y conectores
Get a secure seal at higher pressures.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rates to 220 psi at 77 °F ( 25 °C). Usable from 32 to 212 °F (0 to 100 °C).